Assay Details
Gene Symbol: Caap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643817.1 | XP_007642007.1 | 3 - 4 | 414 | 81 | |
GenBank mRNA | JI891435.1 | - | 3 - 4 | 414 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
