Assay Details
Gene Symbol: Map3k7cl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495510.2 | XP_003495558.1 | 3 - 4 | 236 | 69 | |
XM_007641772.1 | XP_007639962.1 | 5 - 6 | 494 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
