Assay Details
| Gene Symbol: Pin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007640506.1 | XP_007638696.1 | 2 - 3 | 422 | 82 | |
| GenBank mRNA | JI863957.1 | - | 2 - 3 | 422 | 82 | |
More Information
Set Membership: |
|

Pin1
Genomic Map