Assay Details
Gene Symbol: Cdc26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503463.2 | XP_003503511.1 | 2 - 3 | 231 | 77 | |
XM_007644070.1 | XP_007642260.1 | 3 - 4 | 432 | 77 | ||
GenBank mRNA | JI881760.1 | - | 1 - 2 | 122 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
