Assay Details
| Gene Symbol: Fundc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507240.2 | XP_003507288.1 | 3 - 4 | 309 | 67 | |
| XM_007647810.1 | XP_007646000.1 | 4 - 5 | 486 | 67 | ||
| GenBank mRNA | JI872169.1 | - | 3 - 4 | 319 | 67 | |
| JP061153.1 | - | 3 - 4 | 319 | 67 | ||
More Information
Set Membership: |
|

Fundc2
Genomic Map