Assay Details
Gene Symbol: Tmem9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509906.2 | XP_003509954.1 | 4 - 5 | 517 | 58 | |
XM_007650559.1 | XP_007648749.1 | 4 - 5 | 680 | 58 | ||
GenBank mRNA | JI867518.1 | - | 1 - 2 | 139 | 58 | |
JP050073.1 | - | 3 - 4 | 342 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
