Assay Details
| Gene Symbol: LOC100768591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514542.2 | XP_003514590.2 | 7 - 8 | 1109 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100768591| Gene Symbol: LOC100768591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514542.2 | XP_003514590.2 | 7 - 8 | 1109 | 76 | |
Set Membership: |
|