Assay Details
| Gene Symbol: Rsu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505039.2 | XP_003505087.1 | 4 - 5 | 425 | 62 | |
| GenBank mRNA | JI865666.1 | - | 4 - 5 | 427 | 62 | |
| JP049708.1 | - | 4 - 5 | 427 | 62 | ||
More Information
Set Membership: |
|

Rsu1
Genomic Map