Assay Details
Gene Symbol: Mettl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505481.2 | XP_003505529.1 | 3 - 4 | 645 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
