Assay Details
| Gene Symbol: Clec1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509248.2 | XP_003509296.1 | 1 - 2 | 70 | 63 | |
| XM_003509249.2 | XP_003509297.1 | 1 - 2 | 70 | 63 | ||
More Information
Set Membership: |
|

Clec1b
Genomic Map