Assay Details
Gene Symbol: Cxcl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510008.2 | XP_003510056.1 | 2 - 3 | 237 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
