Assay Details
| Gene Symbol: Pgpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007638458.1 | XP_007636648.1 | 6 - 7 | 756 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pgpep1| Gene Symbol: Pgpep1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007638458.1 | XP_007636648.1 | 6 - 7 | 756 | 55 | |
Set Membership: |
|