Assay Details
Gene Symbol: LOC103160220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007637174.1 | XP_007635364.1 | 3 - 4 | 475 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC103160220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007637174.1 | XP_007635364.1 | 3 - 4 | 475 | 97 |
Set Membership: |
![]() ![]() ![]() |