Assay Details
| Gene Symbol: Nenf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652134.1 | XP_007650324.1 | 5 - 6 | 582 | 59 | |
| GenBank mRNA | JI863597.1 | - | 2 - 3 | 180 | 59 | |
More Information
Set Membership: |
|

Nenf
Genomic Map