Assay Details
Gene Symbol: Lgals3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500061.2 | XP_003500109.1 | 3 - 4 | 448 | 65 | |
XM_007640783.1 | XP_007638973.1 | 2 - 3 | 433 | 65 | ||
GenBank mRNA | JI884166.1 | - | 3 - 4 | 417 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
