Assay Details
Gene Symbol: Pddc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509762.2 | XP_003509810.1 | 1 - 2 | 103 | 79 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pddc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509762.2 | XP_003509810.1 | 1 - 2 | 103 | 79 |