Assay Details
| Gene Symbol: Ndufs7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007643194.1 | XP_007641384.1 | 2 - 3 | 165 | 62 | |
| GenBank mRNA | JP039642.1 | - | 3 - 4 | 215 | 62 | |
More Information
Set Membership: |
|

Ndufs7
Genomic Map