Assay Details
| Gene Symbol: Bhmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498697.2 | XP_003498745.1 | 5 - 6 | 594 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bhmt2| Gene Symbol: Bhmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498697.2 | XP_003498745.1 | 5 - 6 | 594 | 63 | |
Set Membership: |
|