Assay Details
Gene Symbol: Tmem30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503191.2 | XP_003503239.1 | 4 - 5 | 553 | 68 | |
GenBank mRNA | JI885848.1 | - | 1 - 2 | 86 | 68 | |
JP043590.1 | - | 4 - 5 | 455 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
