Assay Details
| Gene Symbol: Maf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507669.2 | XP_003507717.1 | 6 - 7 | 977 | 74 | |
| XM_007648242.1 | XP_007646432.1 | 7 - 8 | 1048 | 74 | ||
| XM_007648244.1 | XP_007646434.1 | 6 - 7 | 803 | 74 | ||
| GenBank mRNA | JI882885.1 | - | 6 - 7 | 787 | 74 | |
| JP056432.1 | - | 6 - 7 | 787 | 74 | ||
More Information
Set Membership: |
|

Maf1
Genomic Map