Assay Details
Gene Symbol: LOC100758426 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514977.2 | XP_003515025.2 | 2 - 3 | 928 | 102 | |
XM_007654992.1 | XP_007653182.1 | 3 - 4 | 809 | 102 |

More Information
Set Membership: |
![]() ![]() |
