Assay Details
Gene Symbol: Cnn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643249.1 | XP_007641439.1 | 5 - 6 | 605 | 60 | |
GenBank mRNA | JI879115.1 | - | 2 - 3 | 240 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
