Assay Details
Gene Symbol: Kdelr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648732.1 | XP_007646922.1 | 3 - 4 | 646 | 76 | |
GenBank mRNA | JP046585.1 | - | 3 - 4 | 646 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
