Assay Details
Gene Symbol: Cdrt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649526.1 | XP_007647716.1 | 1 - 2 | 80 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cdrt4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649526.1 | XP_007647716.1 | 1 - 2 | 80 | 59 |
Set Membership: |
![]() ![]() ![]() |