Assay Details
Gene Symbol: Fam92a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648393.1 | XP_007646583.1 | 6 - 7 | 759 | 142 | |
GenBank mRNA | JI865732.1 | - | 6 - 7 | 759 | 142 |

More Information
Set Membership: |
![]() ![]() |
