Assay Details
| Gene Symbol: Pus3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502705.2 | XP_003502753.1 | 2 - 3 | 462 | 72 | |
| XM_007643324.1 | XP_007641514.1 | 2 - 3 | 551 | 72 | ||
| XM_007643325.1 | XP_007641515.1 | 2 - 3 | 615 | 72 | ||
| GenBank mRNA | JI882406.1 | - | 2 - 3 | 440 | 72 | |
| JP060789.1 | - | 2 - 3 | 440 | 72 | ||
More Information
Set Membership: |
|

Pus3
Genomic Map