Assay Details
Gene Symbol: Pawr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508386.2 | XP_003508434.1 | 2 - 3 | 261 | 65 | |
GenBank mRNA | JI873559.1 | - | 1 - 2 | 229 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
