Assay Details
Gene Symbol: Sp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645643.1 | XP_007643833.1 | 1 - 2 | 1372 | 67 | |
XM_007645651.1 | XP_007643841.1 | 1 - 2 | 1372 | 67 | ||
GenBank mRNA | JI871200.1 | - | 1 - 2 | 1234 | 67 | |
JP064027.1 | - | 1 - 2 | 1234 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
