Assay Details
Gene Symbol: Pigz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495389.2 | XP_003495437.1 | 2 - 3 | 570 | 78 | |
XM_007640561.1 | XP_007638751.1 | 2 - 3 | 573 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
