Assay Details
| Gene Symbol: Rpl35a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003495404.2 | XP_003495452.1 | 4 - 5 | 408 | 99 | |
| XM_003495405.2 | XP_003495453.1 | 4 - 5 | 365 | 99 | ||
More Information
Set Membership: |
|

Rpl35a
Genomic Map