Assay Details
| Gene Symbol: Dimt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496211.2 | XP_003496259.1 | 1 - 2 | 215 | 56 | |
| GenBank mRNA | JI862213.1 | - | 1 - 2 | 155 | 56 | |
More Information
Set Membership: |
|

Dimt1
Genomic Map