Assay Details
| Gene Symbol: Elovl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499985.2 | XP_003500033.1 | 5 - 6 | 679 | 62 | |
| GenBank mRNA | JI876606.1 | - | 3 - 4 | 395 | 62 | |
More Information
Set Membership: |
|

Elovl4
Genomic Map