Assay Details
Gene Symbol: Prkra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501021.2 | XP_003501069.1 | 4 - 5 | 558 | 63 | |
GenBank mRNA | JI871158.1 | - | 2 - 3 | 104 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
