Assay Details
Gene Symbol: Slc31a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003503464.2 | XP_003503512.1 | 3 - 4 | 365 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
