Assay Details
| Gene Symbol: LOC100774795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509541.2 | XP_003509589.1 | 3 - 4 | 241 | 67 | |
| GenBank mRNA | JI893073.1 | - | 3 - 4 | 215 | 67 | |
| JP047726.1 | - | 3 - 4 | 215 | 67 | ||
More Information
Set Membership: |
|

LOC100774795
Genomic Map