Assay Details
| Gene Symbol: LOC100774795 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509541.2 | XP_003509589.1 | 6 - 7 | 807 | 85 | |
| GenBank mRNA | JI867644.1 | - | 1 - 2 | 183 | 85 | |
More Information
Set Membership: |
|

LOC100774795
Genomic Map