Assay Details
Gene Symbol: Ggps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509880.2 | XP_003509928.1 | 3 - 4 | 379 | 83 | |
GenBank mRNA | JI895313.1 | - | 1 - 2 | 65 | 83 | |
JP050963.1 | - | 3 - 4 | 379 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
