Assay Details
| Gene Symbol: Cacng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514601.2 | XP_003514649.2 | 1 - 2 | 82 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cacng2| Gene Symbol: Cacng2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003514601.2 | XP_003514649.2 | 1 - 2 | 82 | 69 | |
Set Membership: |
|