Assay Details
Gene Symbol: LOC100758705 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496068.2 | XP_003496116.1 | 2 - 3 | 960 | 89 | |
GenBank mRNA | JI893736.1 | - | 2 - 3 | 502 | 89 | |
JP062624.1 | - | 2 - 3 | 502 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
