Assay Details
| Gene Symbol: Pnpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499565.2 | XP_003499613.1 | 4 - 5 | 560 | 63 | |
| GenBank mRNA | JI879241.1 | - | 4 - 5 | 552 | 63 | |
| JP055242.1 | - | 4 - 5 | 552 | 63 | ||
More Information
Set Membership: |
|

Pnpo
Genomic Map