Assay Details
| Gene Symbol: Cpox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650540.1 | XP_007648730.1 | 4 - 5 | 605 | 66 | |
| GenBank mRNA | JP061328.1 | - | 4 - 5 | 605 | 66 | |
More Information
Set Membership: |
|

Cpox
Genomic Map