Assay Details
| Gene Symbol: Mgll | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500220.2 | XP_003500268.1 | 4 - 5 | 507 | 65 | |
| XM_007640958.1 | XP_007639148.1 | 4 - 5 | 477 | 65 | ||
More Information
Set Membership: |
|

Mgll
Genomic Map