Assay Details
| Gene Symbol: Frrs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500749.2 | XP_003500797.1 | 2 - 3 | 144 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Frrs1l| Gene Symbol: Frrs1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500749.2 | XP_003500797.1 | 2 - 3 | 144 | 74 | |
Set Membership: |
|