Assay Details
| Gene Symbol: S100a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501642.2 | XP_003501690.2 | 3 - 4 | 236 | 78 | |
| XM_007642307.1 | XP_007640497.1 | 2 - 3 | 220 | 78 | ||
| XM_007642308.1 | XP_007640498.1 | 2 - 3 | 331 | 78 | ||
| XM_007642309.1 | XP_007640499.1 | 2 - 3 | 205 | 78 | ||
| XM_007642310.1 | XP_007640500.1 | 2 - 3 | 232 | 78 | ||
| GenBank mRNA | JI881739.1 | - | 2 - 3 | 265 | 78 | |
| JP062297.1 | - | 2 - 3 | 261 | 78 | ||
More Information
Set Membership: |
|

S100a5
Genomic Map