Assay Details
Gene Symbol: Gpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003508297.2 | XP_003508345.1 | 4 - 5 | 951 | 68 | |
GenBank mRNA | JI894144.1 | - | 3 - 4 | 453 | 68 | |
JP049783.1 | - | 3 - 4 | 453 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
