Assay Details
Gene Symbol: Dnase1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650138.1 | XP_007648328.1 | 3 - 4 | 505 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dnase1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650138.1 | XP_007648328.1 | 3 - 4 | 505 | 91 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |