Assay Details
| Gene Symbol: Hexim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504516.2 | XP_003504564.1 | 2 - 3 | 147 | 102 | |
| XM_007645068.1 | XP_007643258.1 | 2 - 3 | 265 | 102 | ||
| XM_007645069.1 | XP_007643259.1 | 2 - 3 | 268 | 102 | ||
| XM_007645070.1 | XP_007643260.1 | 2 - 3 | 230 | 102 | ||
| XM_007645071.1 | XP_007643261.1 | 2 - 3 | 233 | 102 | ||
| XM_007645072.1 | XP_007643262.1 | 2 - 3 | 2024 | 102 | ||
| XM_007645073.1 | XP_007643263.1 | 2 - 3 | 192 | 102 | ||
| XM_007645074.1 | XP_007643264.1 | 2 - 3 | 2027 | 102 | ||
| XM_007645075.1 | XP_007643265.1 | 2 - 3 | 131 | 102 | ||
More Information
Set Membership: |
|

Hexim2
Genomic Map