Assay Details
| Gene Symbol: Plscr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007608471.1 | XP_007606661.1 | 5 - 6 | 1007 | 66 | |
| GenBank mRNA | JI867056.1 | - | 2 - 3 | 292 | 66 | |
More Information
Set Membership: |
|

Plscr3
Genomic Map