Assay Details
Gene Symbol: Ccdc101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514839.2 | XP_003514887.1 | 4 - 5 | 306 | 102 | |
XM_007654883.1 | XP_007653073.1 | 4 - 5 | 522 | 102 | ||
XM_007654884.1 | XP_007653074.1 | 5 - 6 | 661 | 102 | ||
XM_007654885.1 | XP_007653075.1 | 4 - 5 | 307 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
