Assay Details
Gene Symbol: Ccdc101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514839.2 | XP_003514887.1 | 5 - 6 | 377 | 104 | |
XM_007654883.1 | XP_007653073.1 | 5 - 6 | 593 | 104 | ||
XM_007654884.1 | XP_007653074.1 | 6 - 7 | 732 | 104 | ||
XM_007654885.1 | XP_007653075.1 | 5 - 6 | 378 | 104 | ||
GenBank mRNA | JI882703.1 | - | 1 - 2 | 71 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
